missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetic Acid, 30% (w/v) Aqueous Solution, Spectrum™ Chemical
Specifications
Specifications
| Quantity | 1 L |
| Packaging | Amber Glass Bottle |
| Percent Purity | 30% ±1.2% |
| Grade | Chemical Solution |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.