missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[3.3.1]nonane-2,6-dione, 97%, Thermo Scientific™

Catalog No. btb12430da
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB12430DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB12430DA Supplier Thermo Scientific Chemicals Supplier No. BTB12430DA

Chemical Identifiers

CAS 16473-11-3
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00153926
InChI Key QWNPVTXLBMSEPN-UHFFFAOYSA-N
Synonym bicyclo 3.3.1 nonane-2,6-dione, bicyclo 3.3.1 nona-2,6-dione, meerwein's ketone, zlchem 41, acmc-20alon
PubChem CID 140084
IUPAC Name bicyclo[3.3.1]nonane-2,6-dione
SMILES C1CC(=O)C2CCC(=O)C1C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 152.19
Percent Purity 97%
Chemical Name or Material Bicyclo[3.3.1]nonane-2, 6-dione