accessibility menu, dialog, popup

UserName

CAS RN 16473-11-3

IUPAC Name: bicyclo[3.3.1]nonane-2,6-dione
Synonyms: bicyclo[3.3.1]nonane-2,6-dione
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: QWNPVTXLBMSEPN-UHFFFAOYNA-N
SMILES: O=C1CCC2CC1CCC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results