missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[2.2.2]octane-1,4-dicarboxylic Acid 98.0+%, TCI America™

Catalog No. B55951G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B55951G Supplier TCI America Supplier No. B55951G
Only null left

Chemical Identifiers

CAS 711-02-4
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
MDL Number MFCD00498000
InChI Key KVOACUMJSXEJQT-UHFFFAOYSA-N
PubChem CID 607709
IUPAC Name bicyclo[2.2.2]octane-1,4-dicarboxylic acid
SMILES C1CC2(CCC1(CC2)C(=O)O)C(=O)O

Specifications

Melting Point 350°C
Color White
Quantity 1 g
Formula Weight 198.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bicyclo[2.2.2]octane-1,4-dicarboxylic Acid