accessibility menu, dialog, popup

UserName

CAS RN 711-02-4

IUPAC Name: bicyclo[2.2.2]octane-1,4-dicarboxylic acid
Synonyms: bicyclo[2.2.2]octane-1,4-dicarboxylic acid
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: KVOACUMJSXEJQT-UHFFFAOYSA-N
SMILES: OC(=O)C12CCC(CC1)(CC2)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results