missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic Dianhydride 98.0+%, TCI America™

Catalog No. B131725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B131725G 25 g
B1317500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B131725G Supplier TCI America Supplier No. B131725G

Chemical Identifiers

CAS 1719-83-1
Molecular Formula C12H8O6
Molecular Weight (g/mol) 248.19
MDL Number MFCD00082228
InChI Key XLOGCGOPKPCECW-UHFFFAOYSA-N
Synonym bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c:4,5-c' difuran-1,3,5,7 3ah,7ah-tetraone, bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic acid dianhydride, 4,8-etheno-1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, 3a,4,4a,7a,8,8a-hexahydro, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic dianhydride, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic acid dianhydride, 4,10-dioxatetracyclo 5.5.2.0<2,6>.0<8,12> tetradec-13-ene-3,5,9,11-tetraone, 4,10-dioxatetracyclo 5.5.2.0 2 ,?.0?, 1 2 tetradec-13-ene-3,5,9,11-tetrone, bicyclo 2.2.2 oct-7-ene-2,3:5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c
PubChem CID 102679
SMILES C1=CC2C3C(C1C4C2C(=O)OC4=O)C(=O)OC3=O

Specifications

Quantity 25 g
Formula Weight 248.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic Dianhydride