missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic Dianhydride (purified by sublimation) 98.0+%, TCI America™
$102.52 - $445.51
Chemical Identifiers
| CAS | 1719-83-1 |
|---|---|
| Molecular Formula | C12H8O6 |
| Molecular Weight (g/mol) | 248.19 |
| MDL Number | MFCD00082228 |
| InChI Key | XLOGCGOPKPCECW-UHFFFAOYSA-N |
| Synonym | bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c:4,5-c' difuran-1,3,5,7 3ah,7ah-tetraone, bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic acid dianhydride, 4,8-etheno-1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, 3a,4,4a,7a,8,8a-hexahydro, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic dianhydride, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic acid dianhydride, 4,10-dioxatetracyclo 5.5.2.0<2,6>.0<8,12> tetradec-13-ene-3,5,9,11-tetraone, 4,10-dioxatetracyclo 5.5.2.0 2 ,?.0?, 1 2 tetradec-13-ene-3,5,9,11-tetrone, bicyclo 2.2.2 oct-7-ene-2,3:5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c |
| PubChem CID | 102679 |
| SMILES | C1=CC2C3C(C1C4C2C(=O)OC4=O)C(=O)OC3=O |
Chemical Identifiers
| 1719-83-1 | |
| 248.19 | |
| XLOGCGOPKPCECW-UHFFFAOYSA-N | |
| 102679 |
| C12H8O6 | |
| MFCD00082228 | |
| bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c:4,5-c' difuran-1,3,5,7 3ah,7ah-tetraone, bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic acid dianhydride, 4,8-etheno-1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, 3a,4,4a,7a,8,8a-hexahydro, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic dianhydride, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic acid dianhydride, 4,10-dioxatetracyclo 5.5.2.0<2,6>.0<8,12> tetradec-13-ene-3,5,9,11-tetraone, 4,10-dioxatetracyclo 5.5.2.0 2 ,?.0?, 1 2 tetradec-13-ene-3,5,9,11-tetrone, bicyclo 2.2.2 oct-7-ene-2,3:5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c | |
| C1=CC2C3C(C1C4C2C(=O)OC4=O)C(=O)OC3=O |
Specifications
| 1719-83-1 | |
| White-Yellow | |
| MFCD00082228 | |
| bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c:4,5-c' difuran-1,3,5,7 3ah,7ah-tetraone, bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic acid dianhydride, 4,8-etheno-1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, 3a,4,4a,7a,8,8a-hexahydro, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic dianhydride, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic acid dianhydride, 4,10-dioxatetracyclo 5.5.2.0<2,6>.0<8,12> tetradec-13-ene-3,5,9,11-tetraone, 4,10-dioxatetracyclo 5.5.2.0 2 ,?.0?, 1 2 tetradec-13-ene-3,5,9,11-tetrone, bicyclo 2.2.2 oct-7-ene-2,3:5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c | |
| C1=CC2C3C(C1C4C2C(=O)OC4=O)C(=O)OC3=O | |
| 102679 | |
| ≥98.0% (GC,T) | |
| Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic Dianhydride (purified by sublimation) |
| 382°C | |
| C12H8O6 | |
| 5 g | |
| XLOGCGOPKPCECW-UHFFFAOYSA-N | |
| 248.19 | |
| 248.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No