missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 97%

Catalog No. AAA1963214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1963214 25 g
AAA1963222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1963214 Supplier Thermo Scientific Chemicals Supplier No. A1963214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1719-83-1 | C12H8O6 | 248.19 g/mol

Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride is used to prepare the thermo-cured cycloaliphatic polyamic acid and photo-cured cycloaliphatic polyamic ester, which were then converted to cycloaliphatic polyimide film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride is used to prepare the thermo-cured cycloaliphatic polyamic acid and photo-cured cycloaliphatic polyamic ester, which were then converted to cycloaliphatic polyimide film.

Solubility
Soluble in water

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 1719-83-1
Molecular Formula C12H8O6
Molecular Weight (g/mol) 248.19
MDL Number MFCD00082228
InChI Key XLOGCGOPKPCECW-UHFFFAOYSA-N
Synonym bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c:4,5-c' difuran-1,3,5,7 3ah,7ah-tetraone, bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic acid dianhydride, 4,8-etheno-1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, 3a,4,4a,7a,8,8a-hexahydro, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic dianhydride, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic acid dianhydride, 4,10-dioxatetracyclo 5.5.2.0<2,6>.0<8,12> tetradec-13-ene-3,5,9,11-tetraone, 4,10-dioxatetracyclo 5.5.2.0 2 ,?.0?, 1 2 tetradec-13-ene-3,5,9,11-tetrone, bicyclo 2.2.2 oct-7-ene-2,3:5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c
PubChem CID 102679
SMILES C1=CC2C3C(C1C4C2C(=O)OC4=O)C(=O)OC3=O

Specifications

Melting Point ∼360°C
Quantity 25 g
Beilstein 294213
Sensitivity Moisture sensitive
Solubility Information Soluble in water
Formula Weight 248.19
Percent Purity 97%
Chemical Name or Material Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 217-009-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only