missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[2.2.2]oct-5-ene-2-carboxaldehyde 95.0+%, TCI America™

Catalog No. B5279200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5279200MG 200 mg
B52791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5279200MG Supplier TCI America Supplier No. B5279200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38259-00-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
InChI Key KCHFYKPIADOPBJ-UHFFFAOYSA-N
PubChem CID 12493694
IUPAC Name bicyclo[2.2.2]oct-2-ene-5-carbaldehyde
SMILES C1CC2C=CC1CC2C=O

Specifications

Melting Point 13°C
Color Yellow
Boiling Point 73°C
Quantity 200 mg
Formula Weight 136.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Bicyclo[2.2.2]oct-5-ene-2-carboxaldehyde
TSCA No
Recommended Storage Refrigerator