accessibility menu, dialog, popup

UserName

CAS RN 38259-00-6

IUPAC Name: bicyclo[2.2.2]oct-5-ene-2-carbaldehyde
Synonyms: bicyclo[2.2.2]oct-5-ene-2-carbaldehyde
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: KCHFYKPIADOPBJ-UHFFFAOYNA-N
SMILES: O=CC1CC2CCC1C=C2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results