missing translation for 'onlineSavingsMsg'
Learn More

(-)-Bicuculline methiodide, 98%

Catalog No. AAJ67150MF
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67150MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67150MF Supplier Thermo Scientific Chemicals Supplier No. J67150MF

CAS: 40709-69-1 | C21H20INO6 | 509.296 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40709-69-1
Molecular Formula C21H20INO6
Molecular Weight (g/mol) 509.296
MDL Number MFCD00078966
InChI Key HKJKCPKPSSVUHY-GRTNUQQKSA-M
Synonym --bicuculline methiodide, bicuculline methiodide, 1 s ,9 r---bicuculline methiodide, 5s-5-6r-6,8-dihydro-8-oxofuro 3,4-e-1,3-benzodioxol-6-yl-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo 4,5-g isoquinolinium iodide, 1,3-dioxolo 4,5-g isoquinolinium, 5-6r-6,8-dihydro-8-oxofuro 3,4-e-1,3-benzodioxol-6-yl-5,6,7,8-tetrahydro-6,6-dimethyl-, iodide, 5s, 1,3-dioxolo 4,5-g isoquinolinium, 5-6,8-dihydro-8-oxofuro 3,4-e-1,3-benzodioxol-6-yl-5,6,7,8-tetrahydro-6,6-dimethyl-, iodide, r-r*,s*, opera_id_515, --bicucullinemethiodide, 1 s ,9 r---bicuculline methiodide hpce, 5r-5-6s-6,8-dihydro-8-oxofuro 3,4-e-1,3-benzodioxol-6-yl-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo 4,5-g isoquinolinium iodide
PubChem CID 104871
IUPAC Name (6R)-6-[(5S)-6,6-dimethyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium-5-yl]-6H-furo[3,4-g][1,3]benzodioxol-8-one;iodide
SMILES C[N+]1(CCC2=CC3=C(C=C2C1C4C5=C(C6=C(C=C5)OCO6)C(=O)O4)OCO3)C.[I-]

Specifications

Color Yellow
Quantity 50 mg
UN Number UN1544
Beilstein 5419649
Formula Weight 509.3
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (-)-Bicuculline methiodide
Hazard Category H301+H311-H330
Hazard Statement GHS H Statement
H301-H311-H330
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
RTECSNumber JI4904100
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only