missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bicalutamide 98.0+%, TCI America™
$126.76 - $376.54
Chemical Identifiers
| CAS | 90357-06-5 |
|---|---|
| Molecular Formula | C18H14F4N2O4S |
| Molecular Weight (g/mol) | 430.374 |
| MDL Number | MFCD00869971 |
| InChI Key | LKJPYSCBVHEWIU-UHFFFAOYSA-N |
| Synonym | bicalutamide, casodex, cosudex, calutide, bicalutamide cdx, raffolutil, kalumid, casodex tn, bicalutamide usan:inn:ban, bicalutamide casodex |
| PubChem CID | 2375 |
| IUPAC Name | N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenyl)sulfonyl-2-hydroxy-2-methylpropanamide |
| SMILES | CC(CS(=O)(=O)C1=CC=C(C=C1)F)(C(=O)NC2=CC(=C(C=C2)C#N)C(F)(F)F)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B3206200MG
|
TCI America
B3206200MG |
200 mg |
Each for $126.76
|
|
|||||
|
B32061G
|
TCI America
B32061G |
1 g |
Each for $376.54
|
|
|||||
Chemical Identifiers
| 90357-06-5 | |
| 430.374 | |
| LKJPYSCBVHEWIU-UHFFFAOYSA-N | |
| 2375 | |
| CC(CS(=O)(=O)C1=CC=C(C=C1)F)(C(=O)NC2=CC(=C(C=C2)C#N)C(F)(F)F)O |
| C18H14F4N2O4S | |
| MFCD00869971 | |
| bicalutamide, casodex, cosudex, calutide, bicalutamide cdx, raffolutil, kalumid, casodex tn, bicalutamide usan:inn:ban, bicalutamide casodex | |
| N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenyl)sulfonyl-2-hydroxy-2-methylpropanamide |
Specifications
| 90357-06-5 | |
| White | |
| MFCD00869971 | |
| bicalutamide, casodex, cosudex, calutide, bicalutamide cdx, raffolutil, kalumid, casodex tn, bicalutamide usan:inn:ban, bicalutamide casodex | |
| CC(CS(=O)(=O)C1=CC=C(C=C1)F)(C(=O)NC2=CC(=C(C=C2)C#N)C(F)(F)F)O | |
| 430.374 | |
| 430.37 | |
| Crystalline Powder |
| 196°C | |
| C18H14F4N2O4S | |
| 200 mg | |
| LKJPYSCBVHEWIU-UHFFFAOYSA-N | |
| N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenyl)sulfonyl-2-hydroxy-2-methylpropanamide | |
| 2375 | |
| ≥98.0% (HPLC,N) | |
| Bicalutamide |
Safety and Handling
RTECSNumber : TX1413500
TSCA : No