Learn More
Bialaphos sodium, 85%
Bialaphos sodium (bilanafos), CAS # 35597-43-4, is a tripeptide antibiotic used to confirm gene transfer in plants through use of the bar selection gene. | CAS: 71048-99-2 | C11H21N3NaO6P | 345.268 g/mol
$467.81 - $2040.70
Chemical Identifiers
| CAS | 71048-99-2 |
|---|---|
| Molecular Formula | C11H21N3NaO6P |
| Molecular Weight (g/mol) | 345.268 |
| MDL Number | MFCD01674432 |
| InChI Key | RTWIRLHWLMNVCC-WQYNNSOESA-M |
| PubChem CID | 118988468 |
| IUPAC Name | sodium;[(3S)-3-amino-4-[[(2S)-1-[[(1S)-1-carboxyethyl]amino]-1-oxopropan-2-yl]amino]-4-oxobutyl]-methylphosphinate |
| SMILES | CC(C(=O)NC(C)C(=O)O)NC(=O)C(CCP(=O)(C)[O-])N.[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ66802MC
|
Thermo Scientific Chemicals
J66802MC |
100 mg |
Each for $467.81
|
|
|||||
|
AAJ66802ME
|
Thermo Scientific Chemicals
J66802ME |
500 mg |
Each for $2,040.70
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
General Description
- Bialaphos sodium consists of two L-alanyl units attached to L-phosphinothricyl
- Bialaphos is a bacterial metabolite and an antimicrobial
Application
- Bialaphos is commonly used for transformation experiments in some plant species as a selection agent
- Bialaphos sodium can be used as a standard for metabolite and environmental contaminant determination in laboratory settings
Chemical Identifiers
| 71048-99-2 | |
| 345.268 | |
| RTWIRLHWLMNVCC-WQYNNSOESA-M | |
| sodium;[(3S)-3-amino-4-[[(2S)-1-[[(1S)-1-carboxyethyl]amino]-1-oxopropan-2-yl]amino]-4-oxobutyl]-methylphosphinate |
| C11H21N3NaO6P | |
| MFCD01674432 | |
| 118988468 | |
| CC(C(=O)NC(C)C(=O)O)NC(=O)C(CCP(=O)(C)[O-])N.[Na+] |
Specifications
| Bialaphos Sodium | |
| 71048-99-2 | |
| Orange | |
| >110°C (230°F) | |
| MFCD01674432 | |
| 793°C | |
| RTWIRLHWLMNVCC-WQYNNSOESA-M | |
| sodium;[(3S)-3-amino-4-[[(2S)-1-[[(1S)-1-carboxyethyl]amino]-1-oxopropan-2-yl]amino]-4-oxobutyl]-methylphosphinate | |
| 118988468 | |
| 85% |
| Powder | |
| 160°C | |
| 100 mg | |
| C11H21N3NaO6P | |
| UN2811 | |
| Soluble in water | |
| CC(C(=O)NC(C)C(=O)O)NC(=O)C(CCP(=O)(C)[O-])N.[Na+] | |
| 345.268 | |
| 345.26 | |
| Bialaphos sodium |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H301-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber : AY6827000
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only