missing translation for 'onlineSavingsMsg'
Learn More

Bexarotene 96.0+%, TCI America™

Catalog No. B4798100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4798100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4798100MG Supplier TCI America Supplier No. B4798100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 153559-49-0
Molecular Formula C24H28O2
Molecular Weight (g/mol) 348.49
MDL Number MFCD00932428
InChI Key NAVMQTYZDKMPEU-UHFFFAOYSA-N
Synonym bexarotene, targretin, targrexin, targretyn, targret, bexarotenum, bexarotene usan, bexaroteno, bexaroten, targretin-gel
PubChem CID 82146
ChEBI CHEBI:50859
IUPAC Name 4-[1-(3,5,5,8,8-pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethenyl]benzoic acid
SMILES CC1=CC2=C(C=C1C(=C)C1=CC=C(C=C1)C(O)=O)C(C)(C)CCC2(C)C

Specifications

Melting Point 222°C
Color White-Yellow
Quantity 100 mg
Formula Weight 348.49
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bexarotene
RTECSNumber DH6834830
TSCA No
Recommended Storage Freezer (-20 °C)