missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bevacizumab Chimeric Recombinant Mouse Monoclonal Antibody (Bevacizumab)

Mouse Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551653
Description
This antibody binds native and reduced human VEGF (isoform 165).
Bevacizumab (Avastin) is a humanized monoclonal antibody that is approved by the U.S. Food and Drug Administration for the treatment of patients with colon cancer, lung cancer, glioblastoma, renal-cell carcinoma, and age-related macular degeneration. As angiogenesis inhibitor, Bevacizumab blocks angiogenesis by inhibiting vascular endothelial growth factor A (VEGF-A).Chemical Identifiers
| Bevacizumab Chimeric | |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Inhibition Assays, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.02% ProClin 300 | |
| Recombinant human VEGF. | |
| Antibody |
| Bevacizumab | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Human | |
| Avastin | |
| Primary | |
| Protein A |
Specifications
| Bevacizumab Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Mouse | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Flow Cytometry, Inhibition Assays, Immunoprecipitation, Western Blot | |
| Bevacizumab | |
| Unconjugated | |
| Avastin | |
| Recombinant human VEGF. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |