missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Bev-L-Edge™ Slides
Simplify blood smears with beveled edges and chambered corners
Supplier: Fisherbrand™ 2950B2L4600621T
Description
- For smoother, more uniform blood smears, draw beveled edge across a smeared slide at a 45° angle
- Frosted slides are frosted on both sides at one end
Specifications
| Beveled | |
| 1 mm | |
| 3 in. |
| Not Frosted | |
| Chambered | |
| 1 in. |