missing translation for 'onlineSavingsMsg'
Learn More

Betulin, ≥97.5% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111014244
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014244 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014244 Supplier Merck Emd Millipore Supplier No. 9264850MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Betulin is a naturally occurring pentacyclic triterpene alcohol with a lupane skeleton, found in large quantities in the bark of white birch. It may possess anticancer and antiviral properties.

Featured Industries: Food and Beverages

Specifications

CAS 473-98-3
Physical Form Neat
Linear Formula C30H50O2
MDL Number MFCD00016802
Melting Point 256°C to 257°C
Synonym Betulinic alcohol; Betulinol; Betulol; Lup-20(29)-ene-3 beta,28-diol; Trochol
Type Analytical Standard
Quantity 50 mg
Formula Weight 442.72
Percent Purity ≥97.5% (TLC)
Grade Analytical Standard
Show More Show Less