missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Betula pendula BETVIA Chimeric Recombinant Human Monoclonal Antibody (M0418)
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551583
Description
This antibody binds to Bet v 1, in particular to the naturally occurring isoforms Bet v 1.0101 and Bet v 1.0102, but not to Bet v 1.0112.
May be a general steroid carrier protein.Specifications
| Betula pendula BETVIA Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Human | |
| Affinity chromatography | |
| RUO | |
| Plant | |
| Antibody | |
| IgE λ |
| ELISA | |
| M0418 | |
| Unconjugated | |
| allergen Bet v 1-A; Allergen Bet v I-A; BETVI; Major pollen allergen Bet v 1-A | |
| Bet v 1-A, a major allergen of the European white birch. | |
| 50 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |