missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Betamethasone valerate

Catalog No. AC464930010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464930010 1 g
AC464930050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464930010 Supplier Thermo Scientific Chemicals Supplier No. 464930010
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2152-44-5
Molecular Formula C27H37FO6
Molecular Weight (g/mol) 476.59
InChI Key SNHRLVCMMWUAJD-SUYDQAKGSA-N
IUPAC Name (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-10-hydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl pentanoate
SMILES CCCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO

Specifications

Quantity 1 g
Formula Weight 476.58
Chemical Name or Material βmethasone valerate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May damage the unborn child.

Suspected of damaging fertility.

May cause damage to organs through prolonged or repeated exposure.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

EINECSNumber 218-439-3

RUO – Research Use Only