missing translation for 'onlineSavingsMsg'
Learn More

Medisca Betamethasone Sodium Phosphate USP

Catalog No. 14185378
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
10 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185378 100 g
14185282 1 g
14185155 5 g
14185158 500 mg
14185161 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14185378 Supplier Medisca Supplier No. 019505

Betamethasone Sodium Phosphate USP is a white to practically white, odorless powder. It is hygroscopic.

Betamethasone Sodium Phosphate USP is a white to practically white, odorless powder. It is hygroscopic.

Specifications

Chemical Name or Material Betamethasone Sodium Phosphate USP
Packaging Bottle, 500mL, Amber Glass, Wide Mouth Packer, 53-400
CAS 151-73-5
Type Drug Standard
Molecular Weight (g/mol) 516.4072
Quantity 100 g
Percent Purity 97 to 103 %
Grade USP