Learn More
Thermo Scientific™ Betamethasone sodium phosphate
Chemical Identifiers
| CAS | 151-73-5 |
|---|---|
| Molecular Formula | C22H28FNa2O8P |
| Molecular Weight (g/mol) | 516.41 |
| InChI Key | PLCQGRYPOISRTQ-LWCNAHDDSA-L |
| IUPAC Name | disodium (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-1-[2-(phosphonatooxy)acetyl]-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one |
| SMILES | [Na+].[Na+].C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC465740010
|
Thermo Scientific™
465740010 |
1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC465740050
|
Thermo Scientific™
465740050 |
5 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 151-73-5 | |
| 516.41 | |
| disodium (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-1-[2-(phosphonatooxy)acetyl]-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one |
| C22H28FNa2O8P | |
| PLCQGRYPOISRTQ-LWCNAHDDSA-L | |
| [Na+].[Na+].C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP([O-])([O-])=O |
Specifications
| 151-73-5 | |
| 1 g | |
| PLCQGRYPOISRTQ-LWCNAHDDSA-L | |
| disodium (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-1-[2-(phosphonatooxy)acetyl]-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one | |
| 516.41 |
| C22H28FNa2O8P | |
| Solubility aq. soln.: slightly soluble. Other solubilities: soluble in chloroform, methanol | |
| [Na+].[Na+].C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP([O-])([O-])=O | |
| 516.41 | |
| βmethasone sodium phosphate |
Safety and Handling
GHS H Statement Harmful if swallowed.
Suspected of damaging fertility or the unborn child.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 205-797-0
RUO â Research Use Only