missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Betamethasone sodium phosphate
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 206.0°C to 211.0°C |
| Quantity | 1 g |
| Solubility Information | Solubility aq. soln.: slightly soluble. Other solubilities: soluble in chloroform, methanol |
| Formula Weight | 516.41 |
| Chemical Name or Material | βmethasone sodium phosphate |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Suspected of damaging fertility or the unborn child. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection. |
| EINECSNumber | 205-797-0 |
RUO – Research Use Only