missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Betamethasone Recombinant Mouse Monoclonal Antibody (2D5)

Catalog No. MA566616
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA566616 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA566616 Supplier Invitrogen™ Supplier No. MA566616

Mouse Recombinant Monoclonal Antibody

Betamethasone Recombinant Monoclonal Antibody for ELISA

Betamethasone is a potent systemic corticosteroid used primarily to alleviate inflammation in various conditions, including allergic states, dermatologic disorders, gastrointestinal diseases, and hematological disorders. It is often administered in nano-based drug delivery systems to enhance drug release and permeability, reduce dosage, and minimize toxicity. Betamethasone's effectiveness in treating chronic inflammatory skin disorders such as psoriasis and atopic dermatitis is well-documented, with ongoing research focused on improving its delivery and efficacy. The biosimilar market for betamethasone is evolving, with strategies aimed at increasing adoption and addressing safety concerns related to switching from reference biologics to biosimilars. Betamethasone has demonstrated decreased risks for respiratory distress syndrome, intraventricular hemorrhage, and neonatal infections compared to dexamethasone, highlighting its therapeutic advantages.

Specifications

Antigen Betamethasone
Applications ELISA
Classification Recombinant Monoclonal
Clone 2D5
Concentration 10 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Host Species Mouse
Immunogen Conjugate of BET hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less