missing translation for 'onlineSavingsMsg'
Learn More

Betaine Hydrochloride, USP, 98-100.5%, Spectrum™ Chemical

Catalog No. p-7157038
Change view
Click to view available options
Quantity:
100 g
500 g
12 kg
50 kg
Packaging:
Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600934 100 g Poly Bottle
18600935 500 g Poly Bottle
18600937 12 kg Poly Pail
18600938 50 kg Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600934 Supplier Spectrum Chemical Mfg Cor Supplier No. B1104100GM
Only null left

B1104, 590-46-5, C5H11NO2·HCl

Spectrum™ Chemical Betaine Hydrochloride, USP is a vitamin-like substance used to treat hypochlorhydria. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 590-46-5
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00011903
InChI Key HOPSCVCBEOCPJZ-UHFFFAOYSA-N
IUPAC Name (carboxymethyl)trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)CC(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Poly Bottle
Quantity 100 g
Percent Purity 98 to 100.5%
Grade USP