missing translation for 'onlineSavingsMsg'
Learn More

Betaine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7118456
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B045825G 25 g
B0458500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B045825G Supplier TCI America Supplier No. B045825G
Only null left

Chemical Identifiers

CAS 590-46-5
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00011903
InChI Key HOPSCVCBEOCPJZ-UHFFFAOYSA-N
Synonym betaine hydrochloride, betaine chloride, pluchine, acidogeno, acipepsol, achylin, acidin, acinorm, acidol, acidine
PubChem CID 11545
IUPAC Name (carboxymethyl)trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)CC(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 153.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Betaine Hydrochloride
RTECSNumber BP3136000
TSCA Yes