missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betaine Anhydrous 97.0+%, TCI America™
Supplier: TCI America B0455500G
Specifications
| Betaine Anhydrous | |
| 300°C | |
| C5H11NO2 | |
| 500 g | |
| KWIUHFFTVRNATP-UHFFFAOYSA-N | |
| 2-(trimethylazaniumyl)acetate | |
| 247 | |
| 117.15 | |
| Crystalline Powder |
| 107-43-7 | |
| White | |
| MFCD00012123 | |
| betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin | |
| C[N+](C)(C)CC(=O)[O-] | |
| 117.148 | |
| CHEBI:17750 | |
| ≥97.0% (T) |
Chemical Identifiers
| 107-43-7 | |
| 117.148 | |
| KWIUHFFTVRNATP-UHFFFAOYSA-N | |
| 247 | |
| 2-(trimethylazaniumyl)acetate |
| C5H11NO2 | |
| MFCD00012123 | |
| betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin | |
| CHEBI:17750 | |
| C[N+](C)(C)CC(=O)[O-] |
Safety and Handling
EINECSNumber : (2)-1291&(2)-2709
RTECSNumber : DS5900000
TSCA : Yes