missing translation for 'onlineSavingsMsg'
Learn More

Betaine Anhydrous 97.0+%, TCI America™

Catalog No. B045525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B045525G 25 g
B0455100G 100 g
B0455500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B045525G Supplier TCI America Supplier No. B045525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107-43-7
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.148
MDL Number MFCD00012123
InChI Key KWIUHFFTVRNATP-UHFFFAOYSA-N
Synonym betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin
PubChem CID 247
ChEBI CHEBI:17750
IUPAC Name 2-(trimethylazaniumyl)acetate
SMILES C[N+](C)(C)CC(=O)[O-]

Specifications

Melting Point 300°C
Color White
Quantity 25 g
Formula Weight 117.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Betaine Anhydrous
EINECSNumber (2)-1291&(2)-2709
RTECSNumber DS5900000
TSCA Yes