missing translation for 'onlineSavingsMsg'
Learn More

Betaine, 5M Solution, Molecular Biology Grade, Ultrapure

Catalog No. AAJ77507VCR
Change view
Click to view available options
Quantity:
1.5 mL
10 mL
5 x 1.5 mL
100 mL
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ77507VCR 5 x 1.5 mL Plastic bottle
AAJ77507UCR 1.5 mL Plastic bottle
AAJ77507AB 10 mL Plastic bottle
AAJ77507AE 100 mL Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ77507VCR Supplier Thermo Fisher Scientific Supplier No. J77507VCR

CAS: 107-43-7 | C5H11NO2 | 117.148 g/mol

Chemical Identifiers

CAS 107-43-7
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.148
MDL Number MFCD00012123
InChI Key KWIUHFFTVRNATP-UHFFFAOYSA-N
Synonym betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin
PubChem CID 247
ChEBI CHEBI:17750
IUPAC Name 2-(trimethylazaniumyl)acetate
SMILES C[N+](C)(C)CC(=O)[O-]

Specifications

Packaging Plastic bottle
Quantity 5 x 1.5 mL
Beilstein 3537113
Grade Molecular Biology, Ultrapure
Physical Form Solution
Chemical Name or Material Betaine
EINECSNumber 203-490-6
RTECSNumber DS5900000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only