missing translation for 'onlineSavingsMsg'
Learn More

beta-Sitosterol (contains Campesterol) 40.0+%, TCI America™

Catalog No. s004025g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S004025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S004025G Supplier TCI America Supplier No. S004025G
Only null left

Chemical Identifiers

CAS 83-46-5
Molecular Formula C29H50O
Molecular Weight (g/mol) 414.718
MDL Number MFCD00003631
InChI Key KZJWDPNRJALLNS-VJSFXXLFSA-N
Synonym beta-sitosterol, sitosterol, cupreol, quebrachol, 22,23-dihydrostigmasterol, azuprostat, triastonal, cinchol, rhamnol, harzol
PubChem CID 222284
ChEBI CHEBI:27693
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C)C(C)C

Specifications

Melting Point 140°C
Color White
Quantity 25 g
Formula Weight 414.72
Percent Purity ≥40.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material beta-Sitosterol (contains Campesterol)
EINECSNumber (9)-1840
RTECSNumber WJ2600000
TSCA No