Learn More
beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt, 95%
CAS: 2646-71-1 | C21H26N7Na4O17P3 | 833.35 g/mol
Supplier: Thermo Scientific Chemicals J62089MC
Description
NADPH tetra sodium salt is used as an ubiquitous cofactor and biological reducing agent. β-NADPH is a coenzyme found in all living cells and is involved in redox reactions carrying electrons from one reaction to another. It is used as an electron donor, cofactor for many redox enzymes including nitric oxide synthetase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNADPH tetra sodium salt is used as an ubiquitous cofactor and biological reducing agent. β-NADPH is a coenzyme found in all living cells and is involved in redox reactions carrying electrons from one reaction to another. It is used as an electron donor, cofactor for many redox enzymes including nitric oxide synthetase.
Solubility
Soluble in water (50 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Light sensitive. Keep away from strong oxidizing agents.
Specifications
| beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt | |
| White | |
| MFCD10567216 | |
| Light sensitive | |
| Soluble in water (50mg/ml). | |
| [Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O | |
| 833.35 | |
| 833.35 | |
| Powder |
| 2646-71-1 | |
| C21H26N7Na4O17P3 | |
| 100 mg | |
| nadph tetrasodium salt, nadph, tetrasodium salt, coenzyme ii tetrasodium salt reduced form | |
| WYWWVJHQDVCHKF-NRTBITFTNA-J | |
| [[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium | |
| 131673986 | |
| 95% |
Chemical Identifiers
| 2646-71-1 | |
| 833.35 | |
| WYWWVJHQDVCHKF-NRTBITFTNA-J | |
| 131673986 | |
| [Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
| C21H26N7Na4O17P3 | |
| MFCD10567216 | |
| nadph tetrasodium salt, nadph, tetrasodium salt, coenzyme ii tetrasodium salt reduced form | |
| [[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium |
Safety and Handling
EINECSNumber : 220-163-3
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only