accessibility menu, dialog, popup

UserName

N(R)(R)(R)NN(R)OOPPPO(R)(R)(R)(S)ONNOOOOOOOOOOHONH2NH2HOOHNaNaNaNa

CAS RN 2646-71-1

IUPAC Name: tetrasodium [(2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonatooxy)oxolan-2-yl]methyl ({[(2R,3S,4R,5R)-5-(3-carbamoyl-1,4-dihydropyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Synonyms: tetrasodium [(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-(phosphonatooxy)oxolan-2-yl]methyl {[(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxyphosphonate
Molecular Weight (g/mol): 833.35
Molecular Formula: C21H26N7Na4O17P3
InChi Key: WYWWVJHQDVCHKF-NRTBITFTNA-J
SMILES: [Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)