missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Naphthoflavone 99.0+%, TCI America™
$63.38 - $203.17
Chemical Identifiers
| CAS | 6051-87-2 |
|---|---|
| Molecular Formula | C19H12O2 |
| Molecular Weight (g/mol) | 272.303 |
| MDL Number | MFCD00004986 |
| InChI Key | OUGIDAPQYNCXRA-UHFFFAOYSA-N |
| Synonym | beta-naphthoflavone, 5,6-benzoflavone, beta-nf, 3-phenyl-1h-naphtho 2,1-b pyran-1-one, 3-phenyl-1h-benzo f chromen-1-one, 1h-naphtho 2,1-b pyran-1-one, 3-phenyl, unii-1bt0256y8o, 3-phenylbenzo f chromen-1-one, ccris 3262, .beta.-naphthoflavone |
| PubChem CID | 2361 |
| ChEBI | CHEBI:77013 |
| IUPAC Name | 3-phenylbenzo[f]chromen-1-one |
| SMILES | C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC4=CC=CC=C43 |
Chemical Identifiers
| 6051-87-2 | |
| 272.303 | |
| OUGIDAPQYNCXRA-UHFFFAOYSA-N | |
| 2361 | |
| 3-phenylbenzo[f]chromen-1-one |
| C19H12O2 | |
| MFCD00004986 | |
| beta-naphthoflavone, 5,6-benzoflavone, beta-nf, 3-phenyl-1h-naphtho 2,1-b pyran-1-one, 3-phenyl-1h-benzo f chromen-1-one, 1h-naphtho 2,1-b pyran-1-one, 3-phenyl, unii-1bt0256y8o, 3-phenylbenzo f chromen-1-one, ccris 3262, .beta.-naphthoflavone | |
| CHEBI:77013 | |
| C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC4=CC=CC=C43 |
Specifications
| 6051-87-2 | |
| White-Yellow | |
| MFCD00004986 | |
| beta-naphthoflavone, 5,6-benzoflavone, beta-nf, 3-phenyl-1h-naphtho 2,1-b pyran-1-one, 3-phenyl-1h-benzo f chromen-1-one, 1h-naphtho 2,1-b pyran-1-one, 3-phenyl, unii-1bt0256y8o, 3-phenylbenzo f chromen-1-one, ccris 3262, .beta.-naphthoflavone | |
| C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC4=CC=CC=C43 | |
| 272.303 | |
| CHEBI:77013 | |
| ≥99.0% (GC) | |
| beta-Naphthoflavone |
| 165°C | |
| C19H12O2 | |
| 1 g | |
| OUGIDAPQYNCXRA-UHFFFAOYSA-N | |
| 3-phenylbenzo[f]chromen-1-one | |
| 2361 | |
| 272.30 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : QL6200000
TSCA : No