accessibility menu, dialog, popup

UserName

CAS RN 6051-87-2

IUPAC Name: 3-phenyl-1H-benzo[f]chromen-1-one
Synonyms: β-naphthoflavone 5,6-benzoflavone Beta-NF wln: T B666 CV foj ER BNF 5,6 benzoflavone bidd:ER0570 β-NF N3633_sigma
Molecular Weight (g/mol): 272.3
Molecular Formula: C19H12O2
InChi Key: OUGIDAPQYNCXRA-UHFFFAOYSA-N
SMILES: O=C1C=C(OC2=C1C1=CC=CC=C1C=C2)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)