missing translation for 'onlineSavingsMsg'
Learn More

beta-Naphthoflavone 99.0+%, TCI America™

Catalog No. N05181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05181G 1 g
N05185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05181G Supplier TCI America Supplier No. N05181G

Chemical Identifiers

CAS 6051-87-2
Molecular Formula C19H12O2
Molecular Weight (g/mol) 272.303
MDL Number MFCD00004986
InChI Key OUGIDAPQYNCXRA-UHFFFAOYSA-N
Synonym beta-naphthoflavone, 5,6-benzoflavone, beta-nf, 3-phenyl-1h-naphtho 2,1-b pyran-1-one, 3-phenyl-1h-benzo f chromen-1-one, 1h-naphtho 2,1-b pyran-1-one, 3-phenyl, unii-1bt0256y8o, 3-phenylbenzo f chromen-1-one, ccris 3262, .beta.-naphthoflavone
PubChem CID 2361
ChEBI CHEBI:77013
IUPAC Name 3-phenylbenzo[f]chromen-1-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC4=CC=CC=C43

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 1 g
Formula Weight 272.30
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material beta-Naphthoflavone
RTECSNumber QL6200000
TSCA No