missing translation for 'onlineSavingsMsg'
Learn More

beta-Naphthoflavone, 99+%, Thermo Scientific™

$81.75 - $256.11

Chemical Identifiers

CAS 6051-87-2
Molecular Formula C19H12O2
Molecular Weight (g/mol) 272.29
MDL Number MFCD00004986
InChI Key OUGIDAPQYNCXRA-UHFFFAOYSA-N
Synonym beta-naphthoflavone, 5,6-benzoflavone, beta-nf, 3-phenyl-1h-naphtho 2,1-b pyran-1-one, 3-phenyl-1h-benzo f chromen-1-one, 1h-naphtho 2,1-b pyran-1-one, 3-phenyl, unii-1bt0256y8o, 3-phenylbenzo f chromen-1-one, ccris 3262, .beta.-naphthoflavone
PubChem CID 2361
ChEBI CHEBI:77013
IUPAC Name 3-phenylbenzo[f]chromen-1-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC4=CC=CC=C43
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC128170010
View Documents Promotion Details
N/A 1g Glass bottle
Each for $81.75
Only null left
 
AC128170050
View Documents Promotion Details
N/A 5g Glass bottle
Each for $256.11
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6051-87-2
272.29
OUGIDAPQYNCXRA-UHFFFAOYSA-N
2361
3-phenylbenzo[f]chromen-1-one
C19H12O2
MFCD00004986
beta-naphthoflavone, 5,6-benzoflavone, beta-nf, 3-phenyl-1h-naphtho 2,1-b pyran-1-one, 3-phenyl-1h-benzo f chromen-1-one, 1h-naphtho 2,1-b pyran-1-one, 3-phenyl, unii-1bt0256y8o, 3-phenylbenzo f chromen-1-one, ccris 3262, .beta.-naphthoflavone
CHEBI:77013
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC4=CC=CC=C43

Specifications

6051-87-2
100.0
99% min. (HPLC)
MFCD00004986
beta-naphthoflavone, 5,6-benzoflavone, beta-nf, 3-phenyl-1h-naphtho 2,1-b pyran-1-one, 3-phenyl-1h-benzo f chromen-1-one, 1h-naphtho 2,1-b pyran-1-one, 3-phenyl, unii-1bt0256y8o, 3-phenylbenzo f chromen-1-one, ccris 3262, .beta.-naphthoflavone
OUGIDAPQYNCXRA-UHFFFAOYSA-N
3-phenylbenzo[f]chromen-1-one
2361
272.29
β-Naphthoflavone, 99+%
99.0
Authentic
C19H12O2
17, I, 216
Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,chloroform,soluble in sulfuric acid
C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=CC4=CC=CC=C43
272.29
CHEBI:77013
99+%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-958-4

RTECSNumber : QL6200000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only