missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Methyllevulinic Acid 98.0+%, TCI America™
$510.16 - $1726.89
Chemical Identifiers
| CAS | 6628-79-1 |
|---|---|
| Molecular Formula | C6H10O3 |
| Molecular Weight (g/mol) | 130.143 |
| MDL Number | MFCD00143691 |
| InChI Key | NFIWUVRBASXMGK-UHFFFAOYSA-N |
| Synonym | 3-Acetylbutyric Acid, 3-Methyl-4-oxopentanoic Acid |
| PubChem CID | 246523 |
| IUPAC Name | 3-methyl-4-oxopentanoic acid |
| SMILES | CC(CC(=O)O)C(=O)C |
Chemical Identifiers
| 6628-79-1 | |
| 130.143 | |
| NFIWUVRBASXMGK-UHFFFAOYSA-N | |
| 246523 | |
| CC(CC(=O)O)C(=O)C |
| C6H10O3 | |
| MFCD00143691 | |
| 3-Acetylbutyric Acid, 3-Methyl-4-oxopentanoic Acid | |
| 3-methyl-4-oxopentanoic acid |
Specifications
| 6628-79-1 | |
| White-Yellow | |
| C6H10O3 | |
| 5 g | |
| NFIWUVRBASXMGK-UHFFFAOYSA-N | |
| 3-methyl-4-oxopentanoic acid | |
| 246523 | |
| ≥98.0% (GC,T) | |
| beta-Methyllevulinic Acid |
| 32°C | |
| 242°C | |
| MFCD00143691 | |
| 3-Acetylbutyric Acid, 3-Methyl-4-oxopentanoic Acid | |
| CC(CC(=O)O)C(=O)C | |
| 130.143 | |
| 130.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator