missing translation for 'onlineSavingsMsg'
Learn More

beta-Methyllevulinic Acid 98.0+%, TCI America™

Supplier:  TCI America M10575G

 View more versions of this product

Catalog No. M10575G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

beta-Methyllevulinic Acid
32°C
242°C
MFCD00143691
3-Acetylbutyric Acid, 3-Methyl-4-oxopentanoic Acid
CC(CC(=O)O)C(=O)C
130.143
130.14
Crystalline Powder
6628-79-1
White-Yellow
C6H10O3
5 g
NFIWUVRBASXMGK-UHFFFAOYSA-N
3-methyl-4-oxopentanoic acid
246523
≥98.0% (GC,T)

Chemical Identifiers

6628-79-1
130.143
NFIWUVRBASXMGK-UHFFFAOYSA-N
246523
CC(CC(=O)O)C(=O)C
C6H10O3
MFCD00143691
3-Acetylbutyric Acid, 3-Methyl-4-oxopentanoic Acid
3-methyl-4-oxopentanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions