missing translation for 'onlineSavingsMsg'
Learn More

beta-Methylcholine Chloride 98.0+%, TCI America™

Catalog No. M129325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M129325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M129325G Supplier TCI America Supplier No. M129325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2382-43-6
Molecular Formula C6H16ClNO
Molecular Weight (g/mol) 153.65
MDL Number MFCD01669408
InChI Key RUUHDEGJEGHQKL-UHFFFAOYNA-M
Synonym 2-Hydroxypropyltrimethylammonium Chloride
PubChem CID 16941
IUPAC Name (2-hydroxypropyl)trimethylazanium chloride
SMILES [Cl-].CC(O)C[N+](C)(C)C

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 153.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material beta-Methylcholine Chloride
RTECSNumber BR5220000
TSCA Yes