accessibility menu, dialog, popup

UserName

CAS RN 2382-43-6

ClCH3HONCH3CH3H3C

CAS RN 2382-43-6

IUPAC Name: (2-hydroxypropyl)trimethylazanium chloride
Synonyms: β-methylcholine
Molecular Weight (g/mol): 153.65
Molecular Formula: C6H16ClNO
InChi Key: RUUHDEGJEGHQKL-UHFFFAOYNA-M
SMILES: [Cl-].CC(O)C[N+](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results