missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-Lapachone, 98%

Catalog No. AAJ67308LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67308LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67308LB0 Supplier Thermo Scientific Chemicals Supplier No. J67308LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4707-32-8
Molecular Formula C15H14O3

Specifications

Quantity 5 mg
Description Orange
Content And Storage -30°C to -10°C
Formula Weight 242.27
Melting Point 151°C to 153°C
Product Type beta-Lapachone, 98%
Form Solid
Assay 98%

RUO – Research Use Only