CAS RN 4707-32-8
IUPAC Name:
2,2-dimethyl-2H,3H,4H,5H,6H-naphtho[1,2-b]pyran-5,6-dione
Synonyms:
β-lapachone lapachone lapachone, β-
Molecular Weight (g/mol):
242.27
Molecular Formula:
C15H14O3
InChi Key:
QZPQTZZNNJUOLS-UHFFFAOYSA-N
SMILES:
CC1(C)CCC2=C(O1)C1=CC=CC=C1C(=O)C2=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals beta-Lapachone, 98%
CAS: 4707-32-8 Molecular Formula: C15H14O3