accessibility menu, dialog, popup

UserName

HO(R)O(S)O(S)(R)HO(R)OH(R)OHO(R)HO(R)OH(S)HO(R)HO

CAS RN 5965-66-2

IUPAC Name: (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Synonyms: β-D-lactose β-lactose
Molecular Weight (g/mol): 342.3
Molecular Formula: C12H22O11
InChi Key: GUBGYTABKSRVRQ-DCSYEGIMSA-N
SMILES: OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)