missing translation for 'onlineSavingsMsg'
Learn More

beta-Isatoxime 98.0+%, TCI America™

Catalog No. I02735G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I02735G Supplier TCI America Supplier No. I02735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 607-28-3
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00014568
InChI Key LNMAXZZQNSPQSR-UHFFFAOYSA-N
Synonym isatin-3-oxime, 3-hydroxyimino indolin-2-one, 1h-indole-2,3-dione 3-oxime, 3-oximeindole-2,3-dione, isatin beta-oxime, 1h-indole-2,3-dione, 3-oxime, 3-hydroxyiminoindolin-2-one, indole-2,3-dione, 3-oxime, 3e-1h-indole-2,3-dione 3-oxime, 3-hydroxyamino indol-2-one
PubChem CID 5351629
IUPAC Name 3-(hydroxyimino)-2,3-dihydro-1H-indol-2-one
SMILES ON=C1C(=O)NC2=C1C=CC=C2

Specifications

Melting Point 214°C
Quantity 5 g
Formula Weight 162.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material beta-Isatoxime [for Metal Analysis]
RTECSNumber NL7980000
TSCA No