accessibility menu, dialog, popup

UserName

CAS RN 607-28-3

IUPAC Name: 3-(hydroxyimino)-2,3-dihydro-1H-indol-2-one
Synonyms: isatin-3-oxime
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: LNMAXZZQNSPQSR-UHFFFAOYSA-N
SMILES: ON=C1C(=O)NC2=C1C=CC=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results