missing translation for 'onlineSavingsMsg'
Learn More

beta-Hydroxyisovaleric Acid 96.0+%, TCI America™

Supplier:  TCI America H07015G

 View more versions of this product

Catalog No. H07015G


Only null left

Description

Description

Specifications

Specifications

beta-Hydroxyisovaleric Acid
Yellow
MFCD00059081
beta-hydroxyisovaleric acid, 3-hydroxyisovaleric acid, 3-hydroxy-3-methylbutyric acid, hmb-d6, 3-oh-isovaleric acid, 3-hydroxy-isovaleric acid, 3-hydroxy-3-methyl-butanoic acid, butanoic acid, 3-hydroxy-3-methyl, b-hydroxyisovaleric acid, beta-hydroxy-beta-methylbutyrate
CC(C)(O)CC(O)=O
118.13
CHEBI:37084
≥96.0% (GC,T)
625-08-1
C5H10O3
5 g
AXFYFNCPONWUHW-UHFFFAOYSA-N
3-hydroxy-3-methylbutanoic acid
69362
118.13
Liquid

Chemical Identifiers

625-08-1
118.13
AXFYFNCPONWUHW-UHFFFAOYSA-N
69362
3-hydroxy-3-methylbutanoic acid
C5H10O3
MFCD00059081
beta-hydroxyisovaleric acid, 3-hydroxyisovaleric acid, 3-hydroxy-3-methylbutyric acid, hmb-d6, 3-oh-isovaleric acid, 3-hydroxy-isovaleric acid, 3-hydroxy-3-methyl-butanoic acid, butanoic acid, 3-hydroxy-3-methyl, b-hydroxyisovaleric acid, beta-hydroxy-beta-methylbutyrate
CHEBI:37084
CC(C)(O)CC(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1429

TSCA : No

SDS
Documents

Documents

Promotions

Promotions