missing translation for 'onlineSavingsMsg'
Learn More

beta-D-Ribofuranose 1-Acetate 2,3,5-Tribenzoate 98.0+%, TCI America™

Catalog No. p-7135680
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00675G 5 g
R006725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R00675G Supplier TCI America Supplier No. R00675G
Only null left

Chemical Identifiers

CAS 6974-32-9
Molecular Formula C28H24O9
Molecular Weight (g/mol) 504.491
MDL Number MFCD00005357
InChI Key GCZABPLTDYVJMP-CBUXHAPBSA-N
Synonym 1-o-acetyl-2,3,5-tri-o-benzoyl-beta-d-ribofuranose, beta-d-ribofuranose 1-acetate 2,3,5-tribenzoate, 2r,3r,4r,5s-5-acetyloxy-3,4-bis benzoyloxy oxolan-2-yl methyl benzoate, 1-acetyl-2,3,5-tribenzoyl-beta-d-ribofuranose, 1-acetyl-2,3,5-tribenzoy-b-d-ribofuranose, 1-o-acetyl-2,3,5,tri-o-benzyl-beta-d-ribofuranose, 2r,3r,4r,5s-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl methyl benzoate, 2s,3r,4r,5r-2-acetoxy-5-benzoyloxymethyl tetrahydrofuran-3,4-diyl dibenzoate, .beta.-d-ribofuranose, 1-acetate 2,3,5-tribenzoate, 2s,3r,4r,5r-2-acetoxy-5-benzoyloxy methyl tetrahydrofuran-3,4-diyl dibenzoate
PubChem CID 81455
IUPAC Name [(2R,3R,4R,5S)-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl]methyl benzoate
SMILES CC(=O)OC1C(C(C(O1)COC(=O)C2=CC=CC=C2)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 5 g
Formula Weight 504.49
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material beta-D-Ribofuranose 1-Acetate 2,3,5-Tribenzoate
EINECSNumber (9)-0016
TSCA Yes