accessibility menu, dialog, popup

UserName

CH3OOOOOOOOO

CAS RN 6974-32-9

IUPAC Name: [5-(acetyloxy)-3,4-bis(benzoyloxy)oxolan-2-yl]methyl benzoate
Synonyms: [5-(acetyloxy)-3,4-bis(benzoyloxy)oxolan-2-yl]methyl benzoate
Molecular Weight (g/mol): 504.49
Molecular Formula: C28H24O9
InChi Key: GCZABPLTDYVJMP-UHFFFAOYNA-N
SMILES: CC(=O)OC1OC(COC(=O)C2=CC=CC=C2)C(OC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)