missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-D-Ribofuranose 1-Acetate 2,3,5-Tribenzoate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 129°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 504.49 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | beta-D-Ribofuranose 1-Acetate 2,3,5-Tribenzoate |
Safety and Handling
| EINECSNumber | (9)-0016 |
| TSCA | Yes |