missing translation for 'onlineSavingsMsg'
Learn More

beta-D-Glucose (contains alpha-D-Glucose) 85.0+%, TCI America™

Catalog No. G004725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G004725G 25 g
G0047500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G004725G Supplier TCI America Supplier No. G004725G
Only null left

Chemical Identifiers

CAS 492-61-5
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00063989
InChI Key WQZGKKKJIJFFOK-VFUOTHLCSA-N
Synonym beta-d-glucose, beta-d-glucopyranose, glucoside, b-d-glucopyranose, beta-glucose, beta-dextrose, .beta.-d-glucopyranose, b-glucose, b-d-glucose, unii-j4r00m814d
PubChem CID 64689
ChEBI CHEBI:15903
IUPAC Name (2R,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES OC[C@H]1O[C@@H](O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 155°C
Color White
Quantity 25 g
Formula Weight 180.16
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material beta-D-Glucose (contains alpha-D-Glucose)
TSCA Yes
Recommended Storage Refrigerator