missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-D-Glucopyranose 1-Phosphate Disodium Salt 98.0+%, TCI America™
Supplier: TCI America G0339100MG
Specifications
| beta-D-Glucopyranose 1-Phosphate Disodium Salt | |
| White | |
| 100 mg | |
| C(C1C(C(C(C(O1)OP(=O)(O)O)O)O)O)O.[Na].[Na] | |
| 306.114 | |
| 304.10 | |
| Crystalline Powder |
| 83833-15-2 | |
| C6H13Na2O9P | |
| AQMGMJDBUKKFOG-UZUGEDCSSA-N | |
| sodium;[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate | |
| 131885723 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 83833-15-2 | |
| 306.114 | |
| 131885723 | |
| C(C1C(C(C(C(O1)OP(=O)(O)O)O)O)O)O.[Na].[Na] |
| C6H13Na2O9P | |
| AQMGMJDBUKKFOG-UZUGEDCSSA-N | |
| sodium;[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate |
Safety and Handling
EINECSNumber : (8)-0519
TSCA : No
Recommended Storage : Freezer